Learn More
Impacto™ Pearl Leather Series Half-Finger Anti-Impact Gloves
Catalog No. 19031821 Shop All Impacto Protective Products Products
Click to view available options
Size:
Medium
Large
Hand:
Left
Right
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031821 Large Right
19-031811 Medium Left
19-031803 Large Left
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-031821 Supplier Impacto™ Supplier No. 40130110042
This item is not returnable. View return policy
Nylon back with soft pearl leather palm ensures dexterity and comfort. Impact absorbing visco-elastic polymer (VEP) padding in the palm, and adjustable hook and loop wrist closure.
  • Nylon fabric back ensures breathability and comfort
  • Palm is made of soft pearl leather for dexterity and abrasion protection
  • 1/8 in. VEP padding in the palm protects from impact and shock
  • Half-finger style leaves fingers open for tactile feedback and mobility
  • Hook and loop wrist closure
  • Protection against electric, manual, and power tools
  • Ideal for applications such as driving, sports, wheelchairs, and manufacturing
Specifications
Color Black, White
Hand Right
Material Pearl Leather, Nylon
Product Type Fingerless Gloves
Size Large
EN388 Standard EN388:2222
Glove Cuff Style Band Top
Palm Type Padded
Series 401-30