Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Half-Finger Anti-Impact Gloves with Wrist Support

Catalog No. 189994493B Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-4493B Medium
18-999-4493D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-999-4493B Supplier Impacto™ Supplier No. 47530110030
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

A nylon fabric back ensures breathability and comfort, while the attached leather wrist support protects users from repetitive strain injuries.

  • Nylon fabric back ensures breathability and comfort
  • Soft pearl leather cover for dexterity and abrasion protection
  • VEP 1/8 in. padding in the palm, fingers, and thumb web protects from impact and shock
  • Attached leather wrist support prevents over extension and over flexion to protect from repetitive strain injuries
  • Half-finger design for tactile feedback and mobility
  • Provides protection from electric, manual, and power tools
  • Use in applications such as construction, manufacturing, and automotive repairs
  • Gloves are sold in pairs

Specifications

Back Material Nylon
Cuff Material Leather
Color Black, White
Hand Specific
Material Pearl Leather
Product Type Anti-impact Gloves
Size Medium
Finger Style Half
For Use With (Application) Construction, Manufacturing, and Automotive Repairs
Glove Cuff Color White
Series 475-30
Length (English) 8 in.
Length (Metric) 20.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.