Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Leather Palm Tool Grip Gloves

Catalog No. 19031901 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
Hand:
Specific
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031901 Large Specific
19-031904 X-Large Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-031901 Supplier Impacto™ Supplier No. 50120110040
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Nylon gloves are constructed with contoured VEP padding to provide protection from vibration, impact, and abrasion. The grain leather palm is smooth and provides extended wear.

  • Protection from vibration, impact, and abrasion
  • Grain leather palm is soft and durable
  • Fingerless style ensures tactile feedback
  • Use for protection from hand tools and power tools

Specifications

Back Material Nylon Lycra™
Color Blue, Yellow
Hand Specific
Material Grain Leather
Product Type Tool Gloves
Size Large
Finger Style Fingerless
For Use With (Application) Protection from Hand/Power Tools
Glove Cuff Style Open
Series 501-20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.