Learn More
Impacto™ Wrist support with fingerless glove
Catalog No. 19084735 Shop All Impacto Protective Products Products
Click to view available options
Hand:
Left
Right
Specific
Size:
Small
Medium
Large
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand Size
19-084735 Specific X-Large
19-084734 Specific Large
19-084733 Specific Medium
19-062981 Right Small
19-062980 Right Medium
19-076200 Left Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-084735 Supplier Impacto™ Supplier No. 70520010050
Impacto™ 705-20 Fingerless Nylon glove with adjustable wrist support to prevent from RSI
Impacto™ Wrist Support
  • Fingerless nylon Lycra glove with grain leather palm
  • Grain leather wrist support prevents wrist over flexion and over extension to protect from RSI
Specifications
Product Type Impacto wrist support
Length (Metric) 12.7 cm
Length (English) 5 in.
Size X-Large
Hand Specific
Material Leather