Learn More
Impacto™ Air Belt Lumbosacal Lumbar Support
Impacto™ Air Belt Lumbosacral offer extended natural therapy to support the back and help alleviate pain and discomfort
Supplier:  Impacto™ LSXXXL
Catalog No. 19-031703
Only null left
Add to Cart
Description
Description
Impacto™ Lumbosacral Air Belt
  • Inflatable air chambers provide static stretch to relax muscles in spasm
  • Lower portion of contoured bladder system provides support and stability to sacral region
  • Three hold-down straps provide stability and support over lumbosacral area
  • Neoprene stretch fabric in the front and side panels allows four-way freedom of movement and helps keep belt in place
  • Lightweight nylon with moisture-absorbing lining and adjustable hook and loop closure
Specifications
Specifications
3 adjustable straps
3X-Large
Nylon
Back Support
Aspirator w/pressure release valve; Contoured inter connected air chambers; Lightweight, moisture absorbing inner lining; Durable nylon outer cover
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.