Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Glove Liners with VEP Padding

Catalog No. 19088385 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
Hand:
Right
Specific
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-088385 Large Specific
19-031866 Small Right
19-088384 Medium Specific
19-031864 Large Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-088385 Supplier Impacto™ Supplier No. 50100PRLRG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stretch polycotton fabric ensures breathability and comfort, while VEP padding protects palms from impact and shock.

  • 4-way stretch polycotton fabric to ensure breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm
  • Fingerless design for tactile feedback and mobility
  • Designed to be worn under work gloves for an additional level of protection
  • Ideal for protection from electric, manual, and power tools
  • Sold in pairs

Specifications

Certifications/Compliance (Europe) EN 420: 2003 + A1:2009
Color Blue
Hand Specific
Material Polycotton
Product Type Glove Liners
Size Large
EN388 Standard EN388:3231
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Palm Type VEP Padding
Series 501-00
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.