Learn More
Impacto™ Ergomate Tracmates Anti-Slip Safety Soles
Catalog No. 19500181 Shop All Impacto Protective Products Products
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-181 Medium
19-500-180 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-500-181 Supplier Impacto™ Supplier No. TRAC88802M
Impacto™ Ergomates Tracmates are used on any slippery and wet surfaces where there is risk of injury due to falling
Impacto™ Ergomate Tracmates Strap-On Soles
  • Reusable design, ideal for any person who is stripping, washing, or scrubbing floors; food processing, working on slippery surfaces; custodial and janitorial workers, hospitals, kitchens, and health care facilities
  • Not recommended for use on dry floors
  • Gain greater traction when: stripping wax floors, washing and scrubbing surfaces, on slick ceramic tiles, working on slippery concrete, walking in food processing areas, working on wet surfaces
Specifications
Color Black, Green
For Use With (Application) Stripping wax from floor, washing/scrubbing floors, slick kitchen ceramic tiles, slippery concrete, food processing areas, wet surfaces
Material EVA
Size Medium
Toe Material Plain Toe
Gender Unisex
Product Type Safety Soles