Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Tool Grip Gloves with Thumb Web Padding

Catalog No. 19031929 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
X-Large
Hand:
Specific
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031929 X-Large Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-031929 Supplier Impacto™ Supplier No. 50210110051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nylon gloves feature suede leather with VEP padding in the palm, between the thumb, and on the index finger.

  • Nylon fabric offers stretch and breathability
  • Suede leather palm provides abrasion protection
  • Impact and shock absorbing VEP padding in the palm, between the thumb, and index finger
  • VEP padding ensures protection from impact, vibration, and abrasion
  • Thumb web area is protected from cumulative trauma such as bruising from gripping tools

Specifications

Back Material Nylon
Color Blue, Gray
Hand Specific
Material Suede
Product Type Tool Gloves
Size X-Large
For Use With (Application) Handling Tools
Glove Cuff Style Open
Series 502-10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.