Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Half-Finger Trigger Gloves with Wrist Support

Catalog No. 189994597A Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Hand:
Right
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
18-999-4597A Small Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-4597A Supplier Impacto™ Supplier No. 47931110022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed of soft pearl leather on the side of the palm, with a stretchy Nylon Lycra™ back. VEP padding in the palm, trigger finger, and thumb web area provides additional impact protection. Includes padded elastic wrist support with an adjustable hook and loop closure.

  • Soft pearl leather palm offers dexterity and abrasion resistance
  • Nylon Lycra™ back ensures breathability and comfort
  • VEP 1/8 in. padding in the palm, thumb, and index finger to protect from impact and shock
  • Half finger design with extended index finger ensures tactile feedback and mobility
  • Padded elastic wrist support helps prevent over extension and over flexion of the wrist
  • Pull tabs at the fingertips for easy removal
  • Available without leather wrist support as 409-30

Specifications

Back Material Nylon
Cuff Material Elastic
Color Black, White
Hand Right
Material Pearl Leather
Product Type Mechanic's Gloves
Size Small
Finger Style Half
For Use With (Application) Construction, Manufacturing and Maintenance
Glove Cuff Color Black
Glove Cuff Style Hook and Loop
Series 47-931
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.