Learn More
Impacto™ IMPACTOE Steel Toe Cap Overshoes
Catalog No. 19400152 Shop All Impacto Protective Products Products
Click to view available options
Size:
Men: 10 to 11.5, Women: 11.5 to 13
Men: 12 to 12.5
Men: 13 to 14
Men: 6.5 to 7, Women: 8 to 8.5
Men: 8.5 to 9, Women: 10 to 11
Color:
Black
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-400-152 Men: 13 to 14 Black
19-400-147 Men: 6.5 to 7, Women: 8 to 8.5 Black
19-400-149 Men: 8.5 to 9, Women: 10 to 11 Black
19-400-150 Men: 10 to 11.5, Women: 11.5 to 13 Black
19-400-151 Men: 12 to 12.5 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-400-152 Supplier Impacto™ Supplier No. IMPACTOECXXL
Impacto™ Safety Toe Cap protects the toe area and is ideal for any working environment that carries a risk of accidental toe damage or has a steel toe cap requirement.
Impacto™ IMPACTOE Steel toe cap Overshoes
  • Designed to fit tightly over existing shoe
  • Made of vulcanized rubber
  • Flat anti-slip under sole for greasy and oily environment
  • Waterproof - will not harden, crack
  • Acid, oil and animal fat resistant
  • Pull-on-tab and wide shank for true slip-on entry to provide stretch
  • Recommended for use in construction and heavy work environments
  • Transferable from person to person without concern for hygiene or size restraints
  • Color-coded toe for easy identification of sizes
Specifications
Certifications/Compliance CE
Color Black
Material Vulcanized Rubber
Size Men: 13 to 14
Toe Material Steel
Gender Unisex
Waterproof Yes
Product Type IMPACTOE safety toe cap