Learn More
Chemglass Life Sciences Vortexing Marine Style Impeller Blade for 15L Bio Reactors used with 8mm Shaft
SDP
Supplier:  Chemglass Life Sciences CLS138536
Encompass
Marine style impeller blades are available in two styles, Vortexing and Scoping.The 3 through 7 liter sizes fit the 8mm shaft of the agitator assembly, while the 15 liter size fits the 15mm Shaft size.These blades are used primarily for mammalian cell culture work.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.
Catalog No. NC1959051
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.