Learn More
Ace Glass, Inc. Impinger, 125mL, complete with open tube end inlet/outlet, 24/25, 0.05in capillary tip
Supplier: Ace Glass, Inc. 754010
125mL high velocity, air sampling impinger. Supplied with 125mL bottle which accepts 24/25 stopper, stopper with 11mm open tube end inlet/outlet, 8mm O.D. sidearm with bulge, and 0.05 capillary tip.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.