Learn More
Dynalon™ Implosion-proof Desiccators
SCI_ED
Catalog No. S521041 Shop All Dynalab Corporation Products
Catalog No. S521041 Supplier Dynalon™ Supplier No. 243045
Add to Cart
Add to Cart
Includes
Upper and lower sections and perforated polypropylene plate
Upper section is unbreakable, transparent polycarbonate
  • Chemical-resistant polypropylene lower section permits the use of incandescent crucibles (when conventional porcelain plate is substituted for supplied plastic plate)
  • Molded groove in the flange holds the sealing neoprene O-ring
  • Seal is unaffected by low temperature
  • Holds vacuum of 74cm Hg below ambient pressure for 24 hours
Order Info
Retention valve takes 0.6cm I.D. tubing.