Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin4 Antibody [Alexa Fluor Plus 488], R&D Systems™
SDP

Catalog No. RD10021580
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10021580 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10021580 Supplier R&D Systems Supplier No. NBP146166AFP488

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Importin4 Polyclonal antibody specifically detects Importin4 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin),Western Blot (Negative)

Specifications

Antigen Importin4
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)
Classification Polyclonal
Conjugate Alexa Fluor Plus 488
Formulation 50mM Sodium Borate
Gene Alias IMP4B, importin 4, importin-4
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 900 and 950 of human Importin 4 using the numbering given in entry NP_078934.3 (GeneID 79711).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79711
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.