Learn More
Agilent Technologies™ IMT Assay (Anti-CD40) Sample Kit
Catalog No. 8100006
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
81-000-06 1 Kit
1 options
Catalog No. 81-000-06 Supplier Agilent Technologies™ Supplier No. 8100006
Add to Cart
Add to Cart
Liquid tumor killing assay (anti-CD40) sample kit includes tethering reagent (anti-CD40), 10X tethering buffer, and cytolysis reagent.
  • Flexible assays use label free methods for more physiological assay conditions, eliminating labeling or secondary assays
  • Real-time quantitative monitoring of both fast (hours) and slow (days) killing kinetics
  • Sensitive assays capable of evaluating low effector cell to target cell ratios that are physiologically relevant
  • Simple workflow that requires only the addition of effector cells to target cells in the presence or absence of antibodies and minimal sample handling with homogeneous assay
Specifications
Includes 90 μL Tethering Reagent, 10 mL Tethering Buffer, 1.5 mL Cytolysis Reagent
Quantity 1 Kit
Type IMT Assay Kit
System Type CellSensor™