Learn More
Agilent Technologies™ IMT Assay (Anti-CD40) Tethering Kit
Description
- Flexible assays use label free methods for more physiological assay conditions, eliminating labeling or secondary assays
- Real-time quantitative monitoring of both fast (hours) and slow (days) killing kinetics
- Sensitive assays capable of evaluating low effector cell to target cell ratios that are physiologically relevant
- Simple workflow that requires only the addition of effector cells to target cells in the presence or absence of antibodies and minimal sample handling with homogeneous assay
Specifications
Specifications
| Includes | 250 μL Tethering Reagent, 10 mL Tethering Buffer, 1.5 mL Cytolysis Reagent |
| Quantity | 1 Kit |
| Type | IMT Assay Kit |
| System Type | CellSensor™ |
| Druggable Target | Cell-based Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.