Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INAVA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145226 Supplier Thermo Scientific Supplier No. PA5145226
Only null left
Add to Cart
Add to Cart

INAVA Polyclonal Antibody

INAVA Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen INAVA
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C1orf106
Gene Accession No. Q3KP66
Gene Alias 1700034M08Rik; 4933426C09Rik; 5730559C18Rik; AI586180; C1orf106; chromosome 1 open reading frame 106; D1Mgi54; Inava; Innate immunity activator protein; RGD1311892; RIKEN cDNA 5730559C18 gene; similar to hypothetical protein FLJ10901; uncharacterized protein C1orf106; uncharacterized protein C1orf106 homolog
Gene Symbols C1orf106
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 506-595. Recombinant protein control fragment (Product #RP-109992).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.