Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INCA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519229
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519229 Supplier Invitrogen™ Supplier No. PA519229
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

TRUSTED_SUSTAINABILITY

Specifications

Antigen INCA
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CARD17
Gene Accession No. Q5XLA6
Gene Alias CARD17; caspase recruitment domain family member 17; caspase recruitment domain family, member 17; caspase recruitment domain-containing protein 17; Caspase-1 inhibitor INCA; INCA; Inhibitory CARD; inhibitory caspase recruitment domain (CARD) protein; inhibitory caspase recruitment domain protein
Gene Symbols CARD17
Host Species Goat
Immunogen Synthetic peptide sequence (NHLTTQDSQIVLPS) corresponding to the C-terminus amino acids of INCA (aa 97-110).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440068
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.