Learn More
Eurofins Discoverx InCELL Detection Kit, 54 (4 plates), 400/1,600 dp (96-well/384-well). (96-0079S)
Supplier:  Eurofins Discoverx 960079S
Encompass
The InCELL Detection Kit is a detection kit intended for use with InCELL Hunter or Pulse cell lines, allowing measurement of target-engagement based changes in protein stability. This kit contains a lysis buffer and exogenous EA as well as EFC substrate to allow for detection of ProLabel-tagged protein levels in the assayed cells. This kit contains all components required to assay 4 plates (96- or 384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-96-0079s.
Catalog No. NC1528629
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.