Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INCENP Rabbit anti-Human, Mouse, Rat, Polyclonal, liquid, OriGene
SDP

Catalog No. 501684814
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-4814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-4814 Supplier OriGene Supplier No. TA890129
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In mammalian cells, 2 broad groups of centromere-interacting proteins have been described: constitutively binding centromere proteins and ′passenger,′ or transiently interacting, proteins. The constitutive proteins include CENPA (centromere protein A), CENPB, CENPC1, and CENPD. The term ′passenger proteins′ encompasses a broad collection of proteins that localize to the centromere during specific stages of the cell cycle. These include CENPE; MCAK; KID; cytoplasmic dynein; CliPs; and CENPF/mitosin. The inner centromere proteins (INCENPs), the initial members of the passenger protein group, display a broad localization along chromosomes in the early stages of mitosis but gradually become concentrated at centromeres as the cell cycle progresses into mid-metaphase. During telophase, the proteins are located within the midbody in the intercellular bridge, where they are discarded after cytokinesis.

Specifications

Antigen INCENP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene INCENP
Gene Accession No. Q9NQS7, Q9WU62
Gene Alias inner centromere protein
Gene Symbols Incenp
Host Species Rabbit
Immunogen Recombit protein of human INCENP
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16319, 293733, 3619
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.