Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Inclined Plane and Lever Kit
SDPSCI_ED

Catalog No. S83789 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S83789 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S83789 Supplier United Scientific™ Supplier No. UNLVR001
Add to Cart
Add to Cart

Explore basic mechanics concepts with this Inclined Plane and Lever Kit, an all-metal, freestanding apparatus, designed for both durability and ease of use.

  • The plane is wide enough to accommodate standard friction blocks or a Hall's carriage with a smartphone attached.
  • Its walled construction prevents objects from falling off the sides.
  • Quickly adjust the plane's angle and lock it into place with a single knob or flatten the device for easy storage.
  • The included lever features a printed centimeter scale and pre-bored support holes at the center and ends.
  • Binder clips can support hooked or slotted weights, or act as a counterbalance when no weights are attached.

Contents:

  • Aluminum Plane
  • Aluminum Lever, 50 cm Long
  • Support Stand
  • Attachment Rod, with Support Clips
  • 3 Binder Clips

Specifications

Grade 6 to 14
Age 12 to 20
For Use With (Application) Inclined Plane and Lever Demonstrations
Includes Aluminum plane, Aluminum lever, Support Stand, Attachment Rod with support clips, and 3 binder clips
Class Size 1 Lab Group
Product Type Physics Demonstration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.