Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Inclined Plane with Pulley, Roller Only
SDPSCI_ED

Catalog No. S139493 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139493 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139493 Supplier United Scientific™ Supplier No. INPL01R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Roller for Inclined Plane with Pulley.

  • 14 oz. roller that can be used as a vehicle on the plane or as a counterweight
  • For use with inclined plane

Specifications

Grade 5 to 12
Format Kit
Product Type Roller
Includes Roller
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.