Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Inclined Plane with Pulley
SDPSCI_ED

Catalog No. S139492 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139492 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139492 Supplier United Scientific™ Supplier No. INPL01
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sturdy inclined plane kit that includes accessories.

This versatile and tough steel inclined plane includes a graduated protractor and an adjustable pulley.

  • Incline is 4 in. wide and 23 in. long
  • Includes a 14 oz. roller that can be used as a vehicle on the plane or as a counterweight
  • Also includes a pan and cord for masses
  • Includes activity guide

Specifications

Grade 5 to 12
Format Kit
Includes Inclined Plane and Accessories
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.