Learn More
Inconel GE Etch, P4TF8-S12 Class G, Reagents
Inconel GE Etch, P4TF8-S12 Class G, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CI090000500A
| Quantity | 500 mL |
|---|
Chemical Identifiers
| 7732-18-5 , 10025-77-1 | |
| 18.02 | |
| water |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| O |
Specifications
| Brown | |
| H2O | |
| O | |
| 18.02 | |
| Poly Bottle | |
| <1 | |
| Non-sterile | |
| Inconel GE Etch, P4TF8-S12 Class G |
| 7732-18-5 , 10025-77-1 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.2883 | |
| 500 mL | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.