Learn More
Azure Biosystems Incubation Tray
SDP
Catalog No. AC2120AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Dimensions (L x W):
11 x 9 cm
15.5 x 10.5 cm
7 x 5 cm
9 x 6 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
AC2120AZ 7 x 5 cm
AC2122AZ 11 x 9 cm
AC2121AZ 9 x 6 cm
AC2123AZ 15.5 x 10.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC2120AZ Supplier Azure Biosystems Supplier No. AC2120
Add to Cart
Add to Cart
Tray with opaque black lid and bottom for staining and washing light sensitive gels and membranes.
  • Four sizes available
  • These Western blot incubation boxes are suitable for staining and washing gels and membranes. Western blot boxes have smooth interiors to avoid damage to gels and membranes and lids to protect them from dust and debris.
  • Black Western blot boxes are great for light-sensitive applications, like near-infrared fluorescent and chemiluminescent Western blots or protein gels.
Specifications
Color Opaque Black
Type Blot Processing Tray
Dimensions (L x W) 7 x 5 cm