Learn More
Azure Biosystems Incubation Tray
SDP
Catalog No. AC2153AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Dimensions (L x W):
11 x 9 cm
15.5 x 10.5 cm
7 x 5 cm
9 x 6 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
AC2153AZ 15.5 x 10.5 cm
AC2152AZ 11 x 9 cm
AC2151AZ 9 x 6 cm
AC2150AZ 7 x 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC2153AZ Supplier Azure Biosystems Supplier No. AC2153
Add to Cart
Add to Cart
Tray with clear lid and bottom for staining and washing gels and membranes.
  • Four sizes available
  • These Western blot incubation boxes are suitable for staining and washing gels and membranes
  • Western blot boxes have smooth interiors to avoid damage to gels and membranes and lids to protect them from dust and debris
Specifications
Color Clear
Type Blot Processing Tray
Quantity 2 Trays
Dimensions (L x W) 15.5 x 10.5 cm