Learn More
Sartorius Incucyte 3D Nanowell Plate
Supplier:  Sartorius BA04878
Encompass
The Incucyte 3D Nanowell Plates facilitate high-throughput organoid studies by creating a 3D microenvironment that mimics in vivo conditions supporting organoid growth and differentiation. These plates feature an array of hydrogel-printed microwells each optimized to foster uniform and reproducible organoid formation. Additionally each well contains a dedicated pipetting port for seamless medium changes that can be integrated with automated processes thus enhancing efficiency and reproducibility in large-scale screening assays. Together with the Incucyte Live-Cell Analysis System and Incucyte Organoid Analysis Software Module 9600-0034 these plates enable high-throughput long-term imaging and analysis of organoids providing insights into their growth and viability dynamics. This plate also requires systems to be operating on Incucyte Base Software v2024A or later.
Catalog No. 50-289-1654