Learn More
Sartorius Incucyte Clearview Plate
Catalog No. 502891588 Shop All Sartorius Products
Encompass
Track and quantify dynamic cell movement using the Incucyte Clearview 96-well Plate for Chemotaxis. Our optically clear surface enables non-invasive visualization and label-free analysis of cell movement. Incucyte Clearview 96-well Plates have been validated for use with Incucyte Live-Cell Analysis Systems and require the Incucyte Chemotaxis Analysis Software Module. Optical Clarity and Label-Free Analysis The Incucyte Clearview 96-well Plate for Chemotaxis allows you to combine the optical clarity of microfluidic devices with the throughput of transmembrane assays.
Catalog No. 50-289-1588 Supplier Sartorius Supplier No. 4599
Add to Cart
Add to Cart
Request a Quote
Track and quantify dynamic cell movement using the Incucyte Clearview 96-well Plate for Chemotaxis. Our optically clear surface enables non-invasive visualization and label-free analysis of cell movement. Incucyte Clearview 96-well Plates have been validated for use with Incucyte Live-Cell Analysis Systems and require the Incucyte Chemotaxis Analysis Software Module. Optical Clarity and Label-Free Analysis The Incucyte Clearview 96-well Plate for Chemotaxis allows you to combine the optical clarity of microfluidic devices with the throughput of transmembrane assays.