Learn More
Sartorius Incucyte Labeling Kit
Supplier:  Sartorius BA04877
Encompass
Extracellular vesicles EVs are heterogeneous lipid-bound particles 50 – 1000 nm naturally secreted by cells into the extracellular space. Exosomes a subclass of EVs are defined by their size 40 to 160 nm and biogenesis endosomal origin . EVs which transport biomolecules like proteins lipids and nucleic acids are of interest to researchers for diagnostic and therapeutic purposes. The Incucyte Exofluor Green EV Labeling Kit allows for real-time evaluation of cellular EV uptake and is validated for use with the Incucyte Live-Cell Analysis System.
Catalog No. 50-289-1759