Learn More
Sartorius Incucyte Mouse IgG1
Supplier:  Sartorius BA04873
Encompass
Incucyte Mouse IgG1 Fabfluor Antibody Labeling Dye is designed for quick easy labeling of Fc containing test antibodies with a green or red fluorophore. Once labeled the Fabfluor-antibody complex can be used for identification of surface expressed antigens in live cells. In the absence of expressed specific antigen little or no signal is seen on the cells.With the Incucyte integrated analysis software background fluorescence is minimized. This reagent has been validated for use with a number of different antibodies in a range of cell types.
Catalog No. 50-289-1580