Learn More
Sartorius Incucyte Mouse IgG2a
Catalog No. 502891579 Shop All Sartorius Products
Encompass
Incucyte Mouse IgG2a Fabfluor Antibody Labeling Dye is designed for quick easy labeling of Fc containing test antibodies with a green or red fluorophore. Once labeled the Fabfluor-antibody complex can be used for identification of surface expressed antigens in live cells. In the absence of expressed specific antigen little or no signal is seen on the cells. With the Incucyte integrated analysis software background fluorescence is minimized.
Catalog No. 50-289-1579 Supplier Sartorius Supplier No. BA04863
Incucyte Mouse IgG2a Fabfluor Antibody Labeling Dye is designed for quick easy labeling of Fc containing test antibodies with a green or red fluorophore. Once labeled the Fabfluor-antibody complex can be used for identification of surface expressed antigens in live cells. In the absence of expressed specific antigen little or no signal is seen on the cells. With the Incucyte integrated analysis software background fluorescence is minimized.