Learn More
Sartorius Incucyte Neuroactive Kit
Catalog No. 502891607 Shop All Sartorius Products
Encompass
The Incucyte Neuroactive Kit contain cryopreserved vials of rat cortical neurons and astrocytes isolated from embryonic stage E18 Sprague-Dawley rats as well as sufficient Incucyte Neuroburst Orange Lentivirus to perform one 96-well experiment.The Incucyte Neuroburst Orange Lentivirus is a lentiviral based live-cell neuronal labeling reagent driven by a synapsin promoter resulting in the long term expression of a genetically encoded orange fluorescent calcium indicator mRuby-based in neuronal cells.
Catalog No. 50-289-1607 Supplier Sartorius Supplier No. 4761
The Incucyte Neuroactive Kit contain cryopreserved vials of rat cortical neurons and astrocytes isolated from embryonic stage E18 Sprague-Dawley rats as well as sufficient Incucyte Neuroburst Orange Lentivirus to perform one 96-well experiment.The Incucyte Neuroburst Orange Lentivirus is a lentiviral based live-cell neuronal labeling reagent driven by a synapsin promoter resulting in the long term expression of a genetically encoded orange fluorescent calcium indicator mRuby-based in neuronal cells.