Learn More
Sartorius Incucyte Nuclight Rapid NIR Dye for Live-Cell Nuclear Labeling
Catalog No. NC1952323 Shop All Sartorius Products
Encompass
Supplied as a single vial (50 µL), sufficient for 100 to 200 tests in a 96-well plate.
Catalog No. NC1952323 Supplier Sartorius Supplier No. 4804
Supplied as a single vial (50 µL), sufficient for 100 to 200 tests in a 96-well plate.