Learn More
Sartorius Incucyte Nuclight NIR Lentivirus, Efficient Nuclear Labeling, Real-Time Live-Cell Analysis, 0.2 mL
Catalog No. NC2113143 Shop All Sartorius Products
Encompass
The Incucyte Nuclight NIR Lentivirus is a non-toxic lentiviral reagent designed for real-time nuclear labeling of living mammalian cells. It enables live-cell analysis without altering cell function, making it ideal for proliferation and viability studies. With its unique near-infrared fluorescent protein, it provides continuous monitoring capabilities, differentiating it from other products in the market. Its applications include generating stable cell lines and real-time cell counting.
This item is not returnable. View return policy
Catalog No. NC2113143 Supplier Sartorius Supplier No. 4805
The Incucyte Nuclight NIR Lentivirus is a non-toxic lentiviral reagent designed for real-time nuclear labeling of living mammalian cells. It enables live-cell analysis without altering cell function, making it ideal for proliferation and viability studies. With its unique near-infrared fluorescent protein, it provides continuous monitoring capabilities, differentiating it from other products in the market. Its applications include generating stable cell lines and real-time cell counting.