Learn More
Sartorius pHrodo Orange Cell Labeling Kit, Real-Time Phagocytosis Quantification, Compatible With IncuCyte System, 1 Kit
Catalog No. NC2067697 Shop All Sartorius Products
Encompass
The Sartorius IncuCyte pHrodo Orange Cell Labeling Kit provides a reliable solution for quantifying phagocytosis and efferocytosis using live-cell imaging. It features a unique pH-sensitive fluorophore that offers low fluorescence at neutral pH and a significant increase in fluorescence under acidic conditions. This enables real-time evaluation of cellular behaviors affected by various factors, setting it apart from other assays.
Catalog No. NC2067697 Supplier Sartorius Supplier No. 4766
The Sartorius IncuCyte pHrodo Orange Cell Labeling Kit provides a reliable solution for quantifying phagocytosis and efferocytosis using live-cell imaging. It features a unique pH-sensitive fluorophore that offers low fluorescence at neutral pH and a significant increase in fluorescence under acidic conditions. This enables real-time evaluation of cellular behaviors affected by various factors, setting it apart from other assays.