Learn More
Sartorius Incucyte rAstrocytes
Catalog No. 502891711 Shop All Sartorius Products
Encompass
Incucyte rAstrocytes are isolated from the cortex of Sprague Dawley Rats at day 18 of gestation and cryopreserved at 2 000 000 per vial. The ready-to-use vials are intended for use within the Incucyte Live-Cell Analysis System in 96-well co-culture assays with Incucyte rCortical Neurons Cat. No. 4753 for live-cell analysis of Incucyte Neuronal Activity or Incucyte Neurite Analysis.
Catalog No. 50-289-1711 Supplier Sartorius Supplier No. 4586
Incucyte rAstrocytes are isolated from the cortex of Sprague Dawley Rats at day 18 of gestation and cryopreserved at 2 000 000 per vial. The ready-to-use vials are intended for use within the Incucyte Live-Cell Analysis System in 96-well co-culture assays with Incucyte rCortical Neurons Cat. No. 4753 for live-cell analysis of Incucyte Neuronal Activity or Incucyte Neurite Analysis.