Learn More
Sartorius Incucyte rCortical Neurons
Supplier:  Sartorius 4753
Encompass
Incucyte rCortical Neurons are isolated from the cortex of Sprague Dawley Rats at day 18 of gestation and cryopreserved at 2 000 000 per vial. The ready-to-use vials are intended for use within the Incucyte Live-Cell Analysis System in 96-well co-culture assays with Incucyte rAstrocytes Cat. No. 4586 for live-cell analysis of Incucyte Neuronal Activity or Incucyte Neurite Analysis.
Catalog No. 50-289-1770