Learn More
Sartorius Incucyte Reagent Kit
Supplier: Sartorius 4775
The MMP Orange Reagent Kit assesses changes in MMP an indicator of mitochondrial health. The cell-permeable MMP Orange Reagent accumulates in active mitochondria in proportion to the MMP generated by the electrochemical gradient across the mitochondrial membrane.