Learn More
Sartorius Incucyte Reagent Kit
Catalog No. 502891595 Shop All Sartorius Products
Encompass
The Incucyte CytoATP Lentivirus Reagent Kit contains lentiviral based live-cell labeling reagents driven by an EF1-α promotor with puromycin selection resulting in stable expression of a dual fluorescent cpmEGFP and mKOk ATP indicator either ATP binding CytoATP Lentivirus or ATP non-binding control CytoATP Non-binding Control Lentivirus . CytoATP Lentiviruses are highly efficient yet non-disruptive labeling reagents of live cells for the detection and quantification of changes in cytoplasmic ATP in mono- and co-culture cell modelThe Incucyte CytoATP Lentivirus Reagent Kit has been validated for use with the Incucyte Live-Cell Analysis System configured with the Metabolism Optical Module Cat. No. 4820 for measurements of dynamic changes in cytoplasmic ATP. The Incucyte CytoATP Lentivirus Reagent Kit is not compatible with Incucyte instruments configured with Green/Orange/NIR Orange/NIR or Green/Red Optical Modules.
Catalog No. 50-289-1595 Supplier Sartorius Supplier No. 4772
The Incucyte CytoATP Lentivirus Reagent Kit contains lentiviral based live-cell labeling reagents driven by an EF1-α promotor with puromycin selection resulting in stable expression of a dual fluorescent cpmEGFP and mKOk ATP indicator either ATP binding CytoATP Lentivirus or ATP non-binding control CytoATP Non-binding Control Lentivirus . CytoATP Lentiviruses are highly efficient yet non-disruptive labeling reagents of live cells for the detection and quantification of changes in cytoplasmic ATP in mono- and co-culture cell modelThe Incucyte CytoATP Lentivirus Reagent Kit has been validated for use with the Incucyte Live-Cell Analysis System configured with the Metabolism Optical Module Cat. No. 4820 for measurements of dynamic changes in cytoplasmic ATP. The Incucyte CytoATP Lentivirus Reagent Kit is not compatible with Incucyte instruments configured with Green/Orange/NIR Orange/NIR or Green/Red Optical Modules.