Learn More
Sartorius Incucyte S-Series Incustore
Supplier: Sartorius 4689
Ideal if you use your Incucyte intensively as it keeps data on-line and at your fingertips for longer. The RAID 6 configuration of 8 x 6TB hard disks protects all data. If a drive fails you can rest assured that there's no downtime. Simply swap out the drive from the front of the unit and allow the RAID to rebuild.