Learn More
Sartorius INCUCYTE S3 HD/2CLR SYSTEM
Catalog No. 502891707 Shop All Sartorius Products
Encompass
The Incucyte S3 Live-Cell Analysis System is a high-performance instrument designed to image and analyze living cells around the clock for days weeks or months while cells remain undisturbed inside a standard tissue culture incubator. Incucyte S3 is designed for handling higher throughput and accommodates up to six microplates at a time while imaging at 4X 10X or 20X. The Incucyte S3 is a two-color system that comes standard with a Green/Red Optical Module Orange/NIR Optical Module sold separately with HD phase contrast imaging that supports a wide range of 2D and 3D cell culture applications. Multi-user support allows scheduling of experiments at different image acquisition frequencies and magnifications in parallel. 
Catalog No. 50-289-1707 Supplier Sartorius Supplier No. 4647
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Incucyte S3 Live-Cell Analysis System is a high-performance instrument designed to image and analyze living cells around the clock for days weeks or months while cells remain undisturbed inside a standard tissue culture incubator. Incucyte S3 is designed for handling higher throughput and accommodates up to six microplates at a time while imaging at 4X 10X or 20X. The Incucyte S3 is a two-color system that comes standard with a Green/Red Optical Module Orange/NIR Optical Module sold separately with HD phase contrast imaging that supports a wide range of 2D and 3D cell culture applications. Multi-user support allows scheduling of experiments at different image acquisition frequencies and magnifications in parallel. 