Learn More
Sartorius INCUCYTE SX1 HD/G/R SYS PKG
Supplier: Sartorius 4788
The Incucyte SX1 Live-Cell Analysis System is designed to provide smaller labs with a reliable and efficient way to monitor and analyze cell behavior for days weeks or months in real time. The Incucyte SX1 is a two color green/red fluorescence system with HD phase contrast imaging that can support up to two microplates in parallel. It comes standard with a 10X objective and the option to purchase 4X and 20X objectives separately. The system has a robust suite of applications: cell health and viability cell function cell movement and morphology cell monitoring and workflows and assays for 3D models.