Learn More
Sartorius INCUCYTE SX5 HD/3CLR SYS PKG
Catalog No. 502891605 Shop All Sartorius Products
Encompass
The Incucyte SX5 Live-Cell Analysis System is our more advanced offering for live-cell imaging and analysis. It leverages up to five total fluorescence channels up to three simultaneously - including a long wavelength low phototoxicity NIR channel - that makes it ideal for long-term time-lapse experiments. The system offers high-resolution imaging real-time kinetic monitoring and automated analysis capabilities. The system also supports up to six microplates in parallel meaning a wide range of vessels are supported depending on your live-cell imaging and analysis needs.  With a unique moving optical pathway cells remain undisturbed within the physiologically relevant environment of the incubator throughout the course of the assay. Additionally an automated turret boasting 4X/10X/20X objectives makes the Incucyte SX5 our most powerful instrument for multi-user laboratories looking to run multiple assays concurrently.   
Catalog No. 50-289-1605 Supplier Sartorius Supplier No. 4816
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Incucyte SX5 Live-Cell Analysis System is our more advanced offering for live-cell imaging and analysis. It leverages up to five total fluorescence channels up to three simultaneously - including a long wavelength low phototoxicity NIR channel - that makes it ideal for long-term time-lapse experiments. The system offers high-resolution imaging real-time kinetic monitoring and automated analysis capabilities. The system also supports up to six microplates in parallel meaning a wide range of vessels are supported depending on your live-cell imaging and analysis needs.  With a unique moving optical pathway cells remain undisturbed within the physiologically relevant environment of the incubator throughout the course of the assay. Additionally an automated turret boasting 4X/10X/20X objectives makes the Incucyte SX5 our most powerful instrument for multi-user laboratories looking to run multiple assays concurrently.   