Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Indacaterol maleate

Catalog No. AC464650010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464650010 1 g
AC464652500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464650010 Supplier Thermo Scientific Chemicals Supplier No. 464650010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753498-25-8
Molecular Formula C28H32N2O7
Molecular Weight (g/mol) 508.57
MDL Number MFCD20526769,MFCD18782702
InChI Key IREJFXIHXRZFER-UHFFFAOYNA-N
IUPAC Name 5-{2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl}-8-hydroxy-1,2-dihydroquinolin-2-one; but-2-enedioic acid
SMILES OC(=O)C=CC(O)=O.CCC1=C(CC)C=C2CC(CC2=C1)NCC(O)C1=C2C=CC(=O)NC2=C(O)C=C1

Specifications

Melting Point 195.0°C to 197.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso
Formula Weight 508.57
Chemical Name or Material Indacaterol maleate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.