Learn More
Sigma Organic Chemistry Indan | 100ML | 496-11-7 | MFCD00003795
Supplier: Sigma Organic Chemistry I1804100ML

Indan, 100ML
About This Item:
Synonym(s): Hydrindene
Linear Formula: C9H10
Molecular Weight: 118.18
CAS Number: 496-11-7
MDL Number: MFCD00003795
UNSPSC Code: 12352100
Purity: 0.95
Boiling Point: 176 C (lit.)
Melting Point: -51 C (lit.)
Density: 0.965 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.537 (lit.)
Storage: room temp
EINECS Number: 207-814-7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.