Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indan 95.0+%, TCI America™
SDP

Catalog No. I0011100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0011100ML 100 mL
I001125ML 25 mL
I0011500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0011100ML Supplier TCI America Supplier No. I0011100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 496-11-7
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00003795
InChI Key PQNFLJBBNBOBRQ-UHFFFAOYSA-N
Synonym indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
PubChem CID 10326
ChEBI CHEBI:37911
IUPAC Name 2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1

Specifications

Color Yellow
Boiling Point 179°C
Quantity 100 mL
UN Number 3295
Formula Weight 118.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Indan
RTECSNumber NK3750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.