Learn More
Indane, 95%
Description
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.
Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.
Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | -51°C |
| Density | 0.962 |
| Boiling Point | 175°C to 177°C |
| Flash Point | 50°C (122°F) |
| Refractive Index | 1.537 |
| Quantity | 2500 mL |
| UN Number | UN3295 |
| Beilstein | 1904376 |
| Merck Index | 14,4931 |
| Solubility Information | Soluble in ethanol,diethyl ether,chloroform (slightly),and ether. Insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H304 |
| Hazard Statement | GHS H Statement H304-H226 May be fatal if swallowed and enters airways. Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 207-814-7 |
| RTECSNumber | NK3750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.