Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indanthrone, Water Soluble, Spectrum™ Chemical
SDP

Catalog No. 18620913 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-913 25 g
18-620-914 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-913 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I127625GM
Add to Cart
Add to Cart

Synonym: Indanthrene, Vat Blue 4, C.I. Pigment Blue 60, Lutetia Fast Blue RS

Chemical Identifiers

CAS 81-77-6

Specifications

Packaging Amber Glass
Color Dark Blue
Quantity 25 g
Chemical Name or Material Indanthrone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.