Learn More
Market Lab Index Cards for Microscope Slide Box
Supplier:  Market Lab 42272
Encompass
The Index Cards for Microscope Slide Box by MarketLab are 3x5 inch white index cards designed to fit standard microscope slide boxes. They are made from heavy card stock and come in packs of 100 cards. The cards have lines on one side for writing specimen information. They provide an easy way to label microscope slides in slide storage boxes.
Catalog No. NC2889372
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.