Learn More
Abcam Indican Assay Kit, Urine, 8000 to 800,000 nmol/L, 100 Tests
Description
- Sample types: Urine
- Detection range: 8,000 to 800,000 nmol/L
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Indican |
| Detection Location | In-Solution Detection |
| Format | Kit |
| No. of Reactions | 100 Tests |
| Detection Method | Colorimetric |
| Sample Type | Urine |
| Quantity | 100 Tests |
| For Use With (Application) | Functional studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.